Idex® High-Pressure Tees, Crosses, and Manifold with 10-32 Coned Configuration for 1/16" OD Tubing
Combine multiple fluid streams into one under high operating pressures
  • Securely join fluid pathway components Holds tight connection even through vibration
These multiport connectors are designed to maintain a secure joining of tubing or other fluid pathway components in mid- to high-pressure applications.
6,900.00 - ₹70,400.00INR / Each
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Number Of Ports Orifice Dimensions (mm)
Availability Pricing
Idex VHP Tee Assembly, SS, 0.020" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA 0193950 Mfr # U-428
COMPARE
Stainless Steel 3 0.5
₹33,900.00INR / Each
Idex VHP Cross Assembly, SS, 0.020" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA 0193960 Mfr # U-430
COMPARE
Stainless Steel 4 0.5
₹48,100.00INR / Each
Idex VHP Cross Assembly, SS, 0.040" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA 0193962 Mfr # U-431
COMPARE
Stainless Steel 4 1
₹40,700.00INR / Each
Idex High-Pressure Tee Assembly, PEEK, 0.020" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA 0201434 Mfr # P-727
COMPARE
PEEK 3 0.5
₹12,900.00INR / Each
Idex High-Pressure Static Mixing Tee Assembly, PEEK, 0.020" ID, 1/16" OD Tubing, 10-32 Coned, 10 µm SS Frit; 1/EA 0201481 Mfr # U-466S
COMPARE
PEEK 3 0.5
₹45,500.00INR / Each
Idex High-Pressure Manifold Assembly, 7 Port, PEEK, 0.020" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA 0202431 Mfr # P-170
COMPARE
PEEK 7 0.5
₹46,200.00INR / Each
Idex High-Pressure Tee Body, PEEK, 0.020" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA 0202454 Mfr # P-727-01
COMPARE
PEEK 3 0.5
₹6,900.00INR / Each
Idex High-Pressure Tee Assembly, PEEK, 0.050" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA 0202455 Mfr # P-728
COMPARE
PEEK 3 1.25
₹13,800.00INR / Each
Idex High-Pressure Tee Body, PEEK, 0.050" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA 0202456 Mfr # P-728-01
COMPARE
PEEK 3 1.25
₹6,900.00INR / Each
Idex High-Pressure Cross Assembly, PEEK, 0.020" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA 0202457 Mfr # P-729
COMPARE
PEEK 4 0.5
₹18,700.00INR / Each
Idex High-Pressure Cross Body, PEEK, 0.020" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA 0202458 Mfr # P-729-01
COMPARE
PEEK 4 0.5
₹10,200.00INR / Each
Idex VHP Tee Body, SS, 0.020" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA 0202486 Mfr # U-428-01
COMPARE
Stainless Steel 3 0.5
₹30,800.00INR / Each
Idex VHP Tee Body, SS, 0.040" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA 0202487 Mfr # U-429-01
COMPARE
Stainless Steel 3 1
₹35,800.00INR / Each
Idex High-Pressure Static Mixing Tee Assembly, PEEK, 0.020" ID, 1/16" OD Tubing, 10-32 Coned, 10 µm UHMWPE Frit; 1/EA 0202491 Mfr # U-466
COMPARE
PEEK 3 0.5
₹43,600.00INR / Each
Idex High-Pressure Mixing Tee Body, PEEK, 0.020" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA 0202492 Mfr # U-466-02
COMPARE
PEEK 3 0.5
₹70,400.00INR / Each
Idex UltraHigh-Pressure Tee, SS, 0.020" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA 0202507 Mfr # UH-427
COMPARE
Stainless Steel 3 0.5
₹36,600.00INR / Each
Idex Manifold Assembly, PEEK, 9 Port, 1/16" OD Tubing; 1/EA 4271884 Mfr # P-191
COMPARE
PEEK 9 1
₹54,100.00INR / Each
Showing 1- 17 of 17