Idex High-Pressure Static Mixing Tee Assembly, PEEK, 0.020" ID, 1/16" OD Tubing, 10-32 Coned, 10 µm SS Frit; 1/EA
Representative image only.
Idex High-Pressure Static Mixing Tee Assembly, PEEK, 0.020" ID, 1/16" OD Tubing, 10-32 Coned, 10 µm SS Frit; 1/EA
Idex | Mfr: U-466S| Item: 0201481
Combine multiple fluid streams into one under high operating pressures
₹45,500.00INR / Each
₹45,500.00INR / Each
Buy It Together
Current Item
Idex High-Pressure Static Mixing Tee Assembly, PEEK, 0.020" ID, 1/16" OD Tubing, 10-32 Coned, 10 µm SS Frit; 1/EA
Idex High-Pressure Static Mixing Tee Assembly, PEEK, 0.020" ID, 1/16" OD Tubing, 10-32 Coned, 10 µm SS Frit; 1/EA
0201481
₹45,500.00INR / Each
+ + + =
Total Price 100800.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Three fitting assemblies F-300.
Key Features
  • Securely join fluid pathway components Holds tight connection even through vibration
  • PEEK housing and frit
  • 5 frits per pack
  • 1 month warranty
  • 1 month warranty
More About this Item
These multiport connectors are designed to maintain a secure joining of tubing or other fluid pathway components in mid- to high-pressure applications.
1 Month Warranty
Specifications
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Number Of Ports3
  • Thread size UNF(F) (port 1) (InchShort)10-32 Coned
  • Compression Size / Tube Od (Port 1) (in)1/16
  • ColorNatural
  • Max Operating Pressure (PSI)5000
  • Orifice Dimensions (mm)0.5
  • DescriptionHigh-Pressure Static Mixing Tee Assembly, PEEK, 0.020" ID, 1/16" OD Tubing, 10-32 Coned, 10 µm SS Frit; 1/EA