Idex Micro-Metering Valve without Fittings, 10-32 Coned; 1/Ea
Representative image only.
Idex Micro-Metering Valve without Fittings, 10-32 Coned; 1/Ea
Idex | Mfr: P-446NF| Item: 0201692
Excellent for fine control of fluid flow rates
₹20,600.00INR / Each
₹20,600.00INR / Each
Buy It Together
Current Item
Idex Micro-Metering Valve without Fittings, 10-32 Coned; 1/Ea
Idex Micro-Metering Valve without Fittings, 10-32 Coned; 1/Ea
0201692
₹20,600.00INR / Each
+ =
Total Price 33500.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Easily control fluid output for a wide range of low flow applications
  • 10-32 coned configuration
  • 1 year warranty
More About this Item
Micro-metering valves can reduce outgoing flow to as low as 3.5 µL/min. Use valves in peristaltic pump fluid transfer applications, mass spectroscopy, fraction collection, and helium sparging lines. Valves have 0.020" (0.5 mm) thru holes.
Valve has 10-32 coned ports and has 0.020" (0.5 mm) thru holes. Provide your own fitting by matching the port configuration.
Volume listed is maximum volume with valve fully open.
1 Year Warranty
Specifications
  • Valve MaterialPEEK, PTFE
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (InchShort)10-32 coned
  • Volume (µL)7.2
  • ColorBlack
  • DescriptionMicro-Metering Valve Without Fittings, 10-32 Coned; 1/EA