Idex® Manual Chromatography Switching Valves
Easily switch between two fluid paths without disconnecting the lines
  • Enable connections among multiple fluid systems
Manual valves serve well in applications that require operator control or involve lesser frequency of use or injection of smaller sample volumes. Shut off flow during cleaning, column replacement, or other operations without disconnecting the system.
50,400.00 - ₹316,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description Tubing OD (in) Wetted Materials
Availability Pricing
Idex Manual Switching Valve, Double Diagonal, Bulkhead, PEEK/PTFE, 4-Position, 4-Port; 1/EA 0201488 Mfr # V-101D
COMPARE
Manual Switching Valve, Double Diagonal, Bulkhead, PEEK/PTFE, 4-Position, 4-Port; 1/EA 0.0625 PEEK/PTFE
₹50,400.00INR / Each
Idex Manual Switching Valve, Right Angle, Bulkhead, PEEK/PTFE, 4-Position, 4-Port; 1/EA 0201489 Mfr # V-101L
COMPARE
Manual Switching Valve, Right Angle, Bulkhead, PEEK/PTFE, 4-Position, 4-Port; 1/EA 0.0625 PEEK/PTFE
₹53,600.00INR / Each
Idex Manual Switching Valve, Analytical, Large Bore, SS/Vespel®, 2-Position, 6-Port; 1/EA 0201519 Mfr # 7000
COMPARE
Manual Switching Valve, Analytical, Large Bore, SS/Vespel®, 2-Position, 6-Port; 1/EA 0.0625 SS/Vespel
₹259,000.00INR / Each
Idex Manual Switching Valve, Semi-Prep, Large Bore, SS/Vespel®, 2-Position, 6-Port; 1/EA 0201540 Mfr # 7000L
COMPARE
Manual Switching Valve, Semi-Prep, Large Bore, SS/Vespel®, 2-Position, 6-Port; 1/EA 0.0625 SS/Vespel
₹316,000.00INR / Each
Idex Manual Switching Valve, Right Angle, Bulkhead, PEEK, 4-Position, 4-Port; 1/EA 0201578 Mfr # V-101L-60
COMPARE
Manual Switching Valve, Right Angle, Bulkhead, PEEK, 4-Position, 4-Port; 1/EA 0.0625 PEEK
₹53,000.00INR / Each
Idex Manual Switching Valve, Single Tee, Bulkhead, PEEK/PTFE, 4-Position, 4-Port; 1/EA 0201579 Mfr # V-101T
COMPARE
Manual Switching Valve, Single Tee, Bulkhead, PEEK/PTFE, 4-Position, 4-Port; 1/EA 0.0625 PEEK/PTFE
₹51,100.00INR / Each