Idex Manual Switching Valve, Right Angle, Bulkhead, PEEK/PTFE, 4-Position, 4-Port; 1/EA
Representative image only.
Idex Manual Switching Valve, Right Angle, Bulkhead, PEEK/PTFE, 4-Position, 4-Port; 1/EA
Idex | Mfr: V-101L| Item: 0201489
Easily switch between two fluid paths without disconnecting the lines
₹53,600.00INR / Each
₹53,600.00INR / Each
Buy It Together
Current Item
Idex Manual Switching Valve, Right Angle, Bulkhead, PEEK/PTFE, 4-Position, 4-Port; 1/EA
Idex Manual Switching Valve, Right Angle, Bulkhead, PEEK/PTFE, 4-Position, 4-Port; 1/EA
0201489
₹53,600.00INR / Each
+ + =
Total Price 139400.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Flangeless Black PEEK male nuts (4), flangeless natural ETFE ferrules 02019-74 (4), and black port plug 02013-77 (1).
Key Features
  • Enable connections among multiple fluid systems
  • Right angle
  • Bulkhead
  • Swept volume of 6.2 µL
  • PEEK/PTFE construction
  • 1/4-28 flat-bottom ports for 1/16" OD tubing
  • 1 month warranty
More About this Item
Manual valves serve well in applications that require operator control or involve lesser frequency of use or injection of smaller sample volumes. Shut off flow during cleaning, column replacement, or other operations without disconnecting the system.
1 Month Warranty
Specifications
  • Outlet Port1/4-28 flat bottom
  • Inlet Port1/4-28 flat bottom
  • Max Pressure (PSI)500
  • Max Pressure (bar)34
  • Wetted MaterialsPEEK/PTFE
  • Tubing OD (in)0.0625
  • Flow PatternRight angle, four-position
  • DescriptionManual Switching Valve, Right Angle, Bulkhead, PEEK/PTFE, 4-Position, 4-Port; 1/EA