Idex® Low-Pressure Manifolds
Divide a single stream into several or combine several streams into one
  • Manufactured from inert polymers
  • 1/4-28 flat-bottom ports
Made from biocompatible PEEK, they are useful for multiport mixing, multi-well sample injection, or any other operation that requires splitting or combining fluid streams. Use in a variety of low-pressure situations throughout your fluidic system.
34,000.00 - ₹91,400.00INR / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 2) (in) Number Of Ports Orifice Dimensions (mm)
Availability Pricing
Idex Low-Pressure Manifold Assembly, Natural PEEK, 7 Port, 0.040" Bore, 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0201367 Mfr # P-150
COMPARE
1/16 7 1
₹41,000.00INR / Each
Idex Low-Pressure Manifold Assembly, Natural PEEK, 9 Port, 0.062" Bore, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0201368 Mfr # P-190
COMPARE
1/8 9 1.6
₹52,400.00INR / Each
Idex Low-Pressure Manifold Body, Natural PEEK, 7 Port, 0.040" Bore, 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202303 Mfr # P-150-01
COMPARE
1/16 7 1
₹40,600.00INR / Each
Idex Low-Pressure Manifold Assembly, Black PEEK, 7 Port, 0.062" Bore, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202304 Mfr # P-151
COMPARE
1/8 7 1.6
₹40,400.00INR / Each
Idex Low-Pressure Manifold Assembly, Natural PEEK, 6 Port, 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202306 Mfr # P-152
COMPARE
1/16 6 1
₹91,400.00INR / Each
Idex Low-Pressure Manifold Body, Natural PEEK, 6 Port, 0.040" Bore, 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202307 Mfr # P-152-01
COMPARE
1/16 6 1
₹74,900.00INR / Each
Idex Low-Pressure Manifold Assembly, Natural PEEK, 6 Port, 0.062" Bore, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202308 Mfr # P-153
COMPARE
1/8 6 1.6
₹38,300.00INR / Each
Idex Low-Pressure Manifold Assembly, PEEK, 5 Port, 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202310 Mfr # P-154
COMPARE
1/16 5 1
₹46,600.00INR / Each
Idex Low-Pressure Manifold Body, Natural PEEK, 5 Port, 0.040" Bore, 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202311 Mfr # P-154-01
COMPARE
1/16 5 1
₹53,000.00INR / Each
Idex Low-Pressure Manifold Assembly, Natural PEEK, 5 Port, 0.062" Bore, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202312 Mfr # P-155
COMPARE
1/8 5 1.6
₹34,000.00INR / Each
Idex Low-Pressure Manifold Body, Natural PEEK, 5 Port, 0.062" Bore, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202313 Mfr # P-155-01
COMPARE
1/8 5 1.6
₹42,400.00INR / Each
Showing 1- 11 of 11