Idex Manual Switching Valve, Single Tee, Bulkhead, PEEK/PTFE, 4-Position, 4-Port; 1/EA
Representative image only.
Idex Manual Switching Valve, Single Tee, Bulkhead, PEEK/PTFE, 4-Position, 4-Port; 1/EA
Idex | Mfr: V-101T| Item: 0201579
Easily switch between two fluid paths without disconnecting the lines
₹51,100.00INR / Each
₹51,100.00INR / Each
Buy It Together
Current Item
Idex Manual Switching Valve, Single Tee, Bulkhead, PEEK/PTFE, 4-Position, 4-Port; 1/EA
Idex Manual Switching Valve, Single Tee, Bulkhead, PEEK/PTFE, 4-Position, 4-Port; 1/EA
0201579
₹51,100.00INR / Each
+ + =
Total Price 136900.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Flangeless Black PEEK male nuts (4), flangeless natural ETFE ferrules 02019-74 (4), and black port plug 02013-77 (1).
Key Features
  • Enable connections among multiple fluid systems
  • Single tee
  • Bulkhead
  • Swept volume of 9.7 µL
  • 1/4-28 flat-bottom ports for 1/16" OD tubing
  • 1 year warranty
More About this Item
Manual valves serve well in applications that require operator control or involve lesser frequency of use or injection of smaller sample volumes. Shut off flow during cleaning, column replacement, or other operations without disconnecting the system.
1 Year Warranty
Specifications
  • Outlet Port1/4-28 flat bottom
  • Inlet Port1/4-28 flat bottom
  • Max Pressure (PSI)500
  • Max Pressure (bar)34
  • Wetted MaterialsPEEK/PTFE
  • Tubing OD (in)0.0625
  • Flow PatternFour-position
  • DescriptionManual Switching Valve, Single Tee, Bulkhead, PEEK/PTFE, 4-Position, 4-Port; 1/EA