Idex® Low-Pressure Union Assemblies
Make solid connections in low-pressure applications
  • Manufactured from inert polymers
  • 1/4-28 flat-bottom ports
Use straight and elbow assemblies in a variety of low-pressure situations throughout your fluidic system. Assemblies come with fittings.
2,300.00 - ₹10,700.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 2) (in) Fitting Material
Availability Pricing
Idex Low-Pressure Union Assembly, Straight, Natural PEEK, 0.020" Bore, 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA WW-02008-00 Mfr # P-702
COMPARE
1/16 PEEK
₹4,400.00INR / Each
Idex Low-Pressure Union Assembly, Straight, Natural PEEK, 0.050" Bore, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA WW-02008-04 Mfr # P-703
COMPARE
1/8 PEEK
₹4,400.00INR / Each
Idex Low-Pressure Union Assembly, Elbow, Natural PEEK, 0.020" Bore, 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA WW-02023-18 Mfr # P-430
COMPARE
1/16 PEEK
₹10,700.00INR / Each
Idex Low-Pressure Union Assembly, Elbow, Natural PEEK, 0.062" Bore, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA WW-02023-20 Mfr # P-432
COMPARE
1/8 PEEK
₹10,700.00INR / Each
Idex Low-Pressure Union Assembly, True ZDV Straight, Natural ETFE, 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA WW-02023-46 Mfr # P-630
COMPARE
1/16 ETFE
₹2,300.00INR / Each
Idex Low-Pressure Union Assembly, True ZDV Straight, Natural ETFE, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA WW-02023-47 Mfr # P-631
COMPARE
1/8 ETFE
₹2,300.00INR / Each
Idex Low-Pressure Union Assembly, Straight, Natural ETFE, 0.030" Bore, 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA WW-02023-72 Mfr # P-710
COMPARE
1/16 ETFE
₹3,600.00INR / Each
Showing 1- 7 of 7