Idex Low-Pressure Union Assembly, Straight, Natural PEEK, 0.050" Bore, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
Representative image only.
Idex Low-Pressure Union Assembly, Straight, Natural PEEK, 0.050" Bore, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
Idex | Mfr: P-703| Item: 0200804
Make solid connections in low-pressure applications
₹7,800.00INR / Each
₹7,800.00INR / Each
Buy It Together
Current Item
Idex Low-Pressure Union Assembly, Straight, Natural PEEK, 0.050" Bore, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
Idex Low-Pressure Union Assembly, Straight, Natural PEEK, 0.050" Bore, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
0200804
₹7,800.00INR / Each
+ + + =
Total Price 40670.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Four PEEK nut/ETFE ferrule flangeless fittings 02020-99.
Key Features
  • Manufactured from inert polymers
  • 1/4-28 flat-bottom ports
  • Max pressure of 1000 psi (69 bar)
  • 0.050" thru-hole diametr
  • 2.57 µL swept volume
  • PEEK body
More About this Item
Use straight and elbow assemblies in a variety of low-pressure situations throughout your fluidic system. Assemblies come with fittings.
Specifications
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(F) (port 1) (InchShort)1/4-28 Flat Bottom
  • Compression Size / Tube Od (Port 2) (in)1/8
  • Compression Size / Tube Od (Port 2) (mm)3.2
  • ColorNatural
  • Max Temperature (° F)302
  • Max Temperature (° C)150
  • Max Operating Pressure (PSI)1000
  • DescriptionLow-Pressure Union Assembly, Straight, Natural PEEK, 0.050" Bore, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA