Idex Low-Pressure Union Assembly, True ZDV Straight, Natural ETFE, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
Representative image only.
Idex Low-Pressure Union Assembly, True ZDV Straight, Natural ETFE, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
Idex | Mfr: P-631| Item: 0202347
Make solid connections in low-pressure applications
₹4,000.00INR / Each
₹4,000.00INR / Each
Buy It Together
Current Item
Idex Low-Pressure Union Assembly, True ZDV Straight, Natural ETFE, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
Idex Low-Pressure Union Assembly, True ZDV Straight, Natural ETFE, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
0202347
₹4,000.00INR / Each
+ + + =
Total Price 36870.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Two PFA flangeless nuts 02020-37 and two ETFE ferrules 02019-83.
Key Features
  • Manufactured from inert polymers
  • 1/4-28 flat-bottom ports
  • For 1/8" OD tubing
  • Max pressure of 1000 psi (69 bar)
  • True ZDV union
  • ETFE body
  • 1 year warranty
More About this Item
Use straight and elbow assemblies in a variety of low-pressure situations throughout your fluidic system. Assemblies come with fittings.
1 Year Warranty
Specifications
  • Fitting MaterialETFE
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(F) (port 1) (InchShort)1/4-28 Flat Bottom
  • Compression Size / Tube Od (Port 2) (in)1/8
  • Compression Size / Tube Od (Port 2) (mm)3.2
  • ColorNatural
  • Max Operating Pressure (PSI)1000
  • DescriptionLow-Pressure Union Assembly, True ZDV Straight, Natural ETFE, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA