Idex® Inlet Solvent Filter with Stem for Soft Tubing
Protect pump and other system component from damage
  • Stainless steel (SS) stem accepts soft tubing
Filters prevent particulates originating from the sparging system from entering the reservoir, protecting your pump and other system components from damage.
10,300.00 - ₹25,800.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connections Pore Size (µm)
Availability Pricing
Idex Inlet Solvent Filter with Stem, SS, 10 µm, 1/16" ID Tubing; 1/EA 4270100 Mfr # A-302
COMPARE
1/16" OD stem 10
₹13,100.00INR / Each
Idex Inlet Solvent Prep Filter with Stem, SS, 20 µm, 1/16" ID Tubing; 1/EA 4271106 Mfr # A-225
COMPARE
1/16" OD stem 20
₹10,300.00INR / Each
Idex Inlet Solvent Filter with Stem, Precleaned, SS, 10 µm, 1/16" ID Tubing; 1/EA 4271118 Mfr # A-302S
COMPARE
1/16" OD stem 10
₹19,800.00INR / Each
Idex Inlet Solvent Filter with Stem, Bottom-of-the-Bottle, SS, 10 µm, 1/16" ID Tubing; 1/EA 4271119 Mfr # A-309
COMPARE
1/16" OD stem 10
₹11,400.00INR / Each
Idex Inlet Solvent Filter with Stem, SS, 10 µm, 1/8" ID Tubing; 1/EA 4271120 Mfr # A-310
COMPARE
1/8" OD stem 10
₹15,100.00INR / Each
Idex Inlet Solvent Prep Filter with Stem, SS, 10 µm, 1/16" ID Tubing; 1/EA 4271121 Mfr # A-311
COMPARE
1/16" OD stem 10
₹25,800.00INR / Each