IDEX® Hex Head Plugs for 10-32 Coned Ports
Close unused ports to safely store sample
  • Optimize usage of your valves and multiport connectors
Use the plugs to close off unused ports in valves and multiport connectors.
9,800.00 - ₹29,500.00INR / Pkg of 5
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Color Material Description
Availability Pricing
Idex Port Plug, PEEK, Black, 5/16" Hex Head, 10-32 Coned; 5/Pk 0201830 Mfr # 6000-090
COMPARE
Black PEEK Port Plug, 5/16" Hex Head, PEEK, 10-32 Coned; 5/PK
₹29,500.00INR / Pkg of 5
Idex Port Plug, Stainless Steel, 5/16" Hex Head, 10-32 Coned 0201839 Mfr # P-520
COMPARE
Gray SS Port Plug, 5/16" Hex Head, SS, 10-32 Coned; 1/EA
₹9,800.00INR / Each