Idex Port Plug, PEEK, Black, 5/16" Hex Head, 10-32 Coned; 5/Pk
Representative image only.
Idex Port Plug, PEEK, Black, 5/16" Hex Head, 10-32 Coned; 5/Pk
Idex | Mfr: 6000-090| Item: 0201830
Close unused ports to safely store sample
₹29,500.00INR / Pkg of 5
₹29,500.00INR / Pkg of 5
Product Description
Key Features
  • Optimize usage of your valves and multiport connectors
  • 5/16" hex head
  • PEEK construction
  • 1 year warranty
More About this Item
Use the plugs to close off unused ports in valves and multiport connectors.
1 Year Warranty
Specifications
  • MaterialPEEK
  • ColorBlack
  • Compatible With10-32 coned port
  • DescriptionPort Plug, 5/16" Hex Head, PEEK, 10-32 Coned; 5/PK