Idex Port Plug, Stainless Steel, 5/16" Hex Head, 10-32 Coned
Representative image only.
Idex Port Plug, Stainless Steel, 5/16" Hex Head, 10-32 Coned
Idex | Mfr: P-520| Item: 0201839
Close unused ports to safely store sample
₹9,800.00INR / Each
₹9,800.00INR / Each
Product Description
Key Features
  • Optimize usage of your valves and multiport connectors
  • 5/16" hex head
  • SS construction
  • 1 year warranty
More About this Item
Use the plugs to close off unused ports in valves and multiport connectors.
1 Year Warranty
Specifications
  • MaterialSS
  • ColorGray
  • Compatible With10-32 coned port
  • DescriptionPort Plug, 5/16" Hex Head, SS, 10-32 Coned; 1/EA