Idex® Flangeless Standard Knurl Fitting Assemblies, 10-32 Flat-Bottom Connection for 1/16" OD Tubing
Eliminate the need to flange tubing
  • Removable and reusable components save you time and money
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components.
1,700.00 - ₹15,800.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Compression Size / Tube Od (Port 1) (in) Fitting Material
Availability Pricing
Idex Flangeless Fitting, Standard Knurl, PEEK, 1/16" OD Tubing, 10-32 Flat-Bottom; 1/EA 0202055 Mfr # XLT-111
COMPARE
Natural 1/16 PEEK
₹1,700.00INR / Each
Idex Flangeless Fitting, Standard Knurl, PEEK, 1/16" OD Tubing, 10-32 Flat-Bottom; 10/PK 0202056 Mfr # XLT-111X
COMPARE
Natural 1/16 PEEK
₹15,800.00INR / Pkg of 10