Idex Flangeless Fitting, Standard Knurl, PEEK, 1/16" OD Tubing, 10-32 Flat-Bottom; 1/EA
Representative image only.
Idex Flangeless Fitting, Standard Knurl, PEEK, 1/16" OD Tubing, 10-32 Flat-Bottom; 1/EA
Idex | Mfr: XLT-111| Item: 0202055
Eliminate the need to flange tubing
₹1,700.00INR / Each
₹1,700.00INR / Each
Buy It Together
Current Item
Idex Flangeless Fitting, Standard Knurl, PEEK, 1/16" OD Tubing, 10-32 Flat-Bottom; 1/EA
Idex Flangeless Fitting, Standard Knurl, PEEK, 1/16" OD Tubing, 10-32 Flat-Bottom; 1/EA
0202055
₹1,700.00INR / Each
+ =
Total Price 2190.00 ADD 2 ITEMS TO CART
Product Description
Includes
Natural ETFE ferrule 02019-74.
Key Features
  • Removable and reusable components save you time and money
  • Natural PEEK
  • 1/EA
  • 1 year warranty
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components.
1 Year Warranty
Specifications
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (InchShort)10-32 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/16