Idex Flangeless Fitting, Standard Knurl, PEEK, 1/16" OD Tubing, 10-32 Flat-Bottom; 10/PK
Representative image only.
Idex Flangeless Fitting, Standard Knurl, PEEK, 1/16" OD Tubing, 10-32 Flat-Bottom; 10/PK
Idex | Mfr: XLT-111X| Item: 0202056
Eliminate the need to flange tubing
₹15,800.00INR / Pkg of 10
₹15,800.00INR / Pkg of 10
Buy It Together
Current Item
Idex Flangeless Fitting, Standard Knurl, PEEK, 1/16" OD Tubing, 10-32 Flat-Bottom; 10/PK
Idex Flangeless Fitting, Standard Knurl, PEEK, 1/16" OD Tubing, 10-32 Flat-Bottom; 10/PK
0202056
₹15,800.00INR / Pkg of 10
+ =
Total Price 16290.00 ADD 2 ITEMS TO CART
Product Description
Includes
Natural ETFE ferrules 02019-74.
Key Features
  • Removable and reusable components save you time and money
  • Natural PEEK
  • 10/PK
  • 1 year warranty
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components.
1 Year Warranty
Specifications
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (InchShort)10-32 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/16