Idex® Flangeless Specialty Head Male Nuts, 1/4-28 Flat-Bottom Connection
Excellent for tight-space applications
  • Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
  • Available in headless, FlashNut™, or short head configurations
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Select from various ferrule materials to suit your application.
1,100.00 - ₹4,500.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Fitting Material Description
Availability Pricing
Idex Flangeless Male Nut, FlushNut™, SS, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0201921 Mfr # F-358
COMPARE
1/16 SS
₹4,500.00INR / Each
Idex Flangeless Male Nut, FlushNut™, SS, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0201922 Mfr # F-368
COMPARE
1/8 SS
₹4,300.00INR / Each
Idex Flangeless Male Nut, Headless, Natural PPS, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0201980 Mfr # P-286
COMPARE
1/16 PPS
₹1,100.00INR / Each