Idex Flangeless Male Nut, Headless, Natural PPS, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
Representative image only.
Idex Flangeless Male Nut, Headless, Natural PPS, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
Idex | Mfr: P-286| Item: 0201980
Excellent for tight-space applications
₹1,100.00INR / Each
₹1,100.00INR / Each
Buy It Together
Current Item
Idex Flangeless Male Nut, Headless, Natural PPS, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
Idex Flangeless Male Nut, Headless, Natural PPS, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
0201980
₹1,100.00INR / Each
+ =
Total Price 1580.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
  • Available in headless, FlashNut™, or short head configurations
  • Headless
  • Natural PPS
  • For 1/16" OD tubing
  • 1/EA
  • 1 year warranty
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Select from various ferrule materials to suit your application.
1 Year Warranty
Specifications
  • Fitting MaterialPPS
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (InchShort)1/4-28 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/16