Idex Flangeless Male Nut, FlushNut™, SS, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
Representative image only.
Idex Flangeless Male Nut, FlushNut™, SS, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
Idex | Mfr: F-368| Item: 0201922
Excellent for tight-space applications
₹4,300.00INR / Each
₹4,300.00INR / Each
Product Description
Key Features
  • Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
  • Available in headless, FlashNut™, or short head configurations
  • FlushNut™
  • Stainless steel
  • For 1/8" OD tubing
  • 1/EA
  • 1 year warranty
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Select from various ferrule materials to suit your application.
1 Year Warranty
Specifications
  • Fitting MaterialSS
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (InchShort)1/4-28 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/8