HTL Discovery Comfort Pipettors
Comfort, safety, precision and value make this variable-volume pipette series quite a discovery
  • Volume locking prevents accidental counter shift
  • Ergonomic design, low pipetting forces help curb repetitive stress injury
  • Fully autoclavable
  • Single-, 8- and 12-channel versions available
Meticulously designed to ensure your comfort, this UV-resistant pipettor is easy-to-use and calibrate. Discover a 360° rotating manifold, tip ejection with minimal effort, perfect tip sealing and excellent precision. Multichannel models feature a revolutionary suspension system which allows the shafts to retract slightly when they are pressed against a row of pipette tips.
Call For Price
27 variations of this product are available.
See All Product Options Below
27 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Code Max Volume (µL) Number Of Channels
Availability Pricing
HTL 4040 Discovery Comfort DV250 Single Channel Pipettor with Gray Shaft, Green; 50 to 250 µL 2455135 Mfr # 4040
COMPARE
Green 250 1
Call For Price
HTL 4041 Discovery Comfort DV2 Single Channel Pipettor with Gray Shaft, Red; 0.2 to 2 µL 2455136 Mfr # 4041
COMPARE
Red 2 1
Call For Price
HTL 4042 Discovery Comfort DV10 Single Channel Pipettor with Gray Shaft, Red; 0.5 to 10 µL 2455137 Mfr # 4042
COMPARE
Red 10 1
Call For Price
HTL 4043 Discovery Comfort DV20 Single Channel Pipettor with Gray Shaft, Yellow; 2 to 20 µL 2455138 Mfr # 4043
COMPARE
Yellow 20 1
Call For Price
HTL 4045 Discovery Comfort DV200 Single Channel Pipettor with Gray Shaft, Yellow; 20 to 200 µL 2455139 Mfr # 4045
COMPARE
Yellow 200 1
Call For Price
HTL 4046 Discovery Comfort DV1000 Single Channel Pipettor with Gray Shaft, Blue; 100 to 1000 µL 2455140 Mfr # 4046
COMPARE
Blue 1000 1
Call For Price
HTL 4047 Discovery Comfort DV50 Single Channel Pipettor with Gray Shaft, Yellow; 5 to 50 µL 2455141 Mfr # 4047
COMPARE
Yellow 50 1
Call For Price
HTL 4048 Discovery Comfort DV5000 Single Channel Pipettor with Gray Shaft, White; 500 to 5000 µL 2455142 Mfr # 4048
COMPARE
White 5000 1
Call For Price
HTL 4049 Discovery Comfort DV10000 Single Channel Pipettor with Gray Shaft, White; 1 to 10 mL 2455143 Mfr # 4049
COMPARE
White 10 1
Call For Price
HTL 4050 Discovery Comfort D250 Single Channel Pipettor with Colored Shaft, Green; 50 to 250 µL 2455144 Mfr # 4050
COMPARE
Green 250 1
Call For Price
HTL 4051 Discovery Comfort D2 Single Channel Pipettor with Colored Shaft, Red; 0.2 to 2 µL 2455145 Mfr # 4051
COMPARE
Red 2 1
Call For Price
HTL 4052 Discovery Comfort D10 Single Channel Pipettor with Colored Shaft, Red; 0.5 to 10 µL 2455146 Mfr # 4052
COMPARE
Red 10 1
Call For Price
HTL 4053 Discovery Comfort D20 Single Channel Pipettor with Colored Shaft, Yellow; 2 to 20 µL 2455147 Mfr # 4053
COMPARE
Yellow 20 1
Call For Price
HTL 4054 Discovery Comfort D100 Single Channel Pipettor with Colored Shaft, Yellow; 10 to 100 µL 2455148 Mfr # 4054
COMPARE
Yellow 100 1
Call For Price
HTL 4055 Discovery Comfort D200 Single Channel Pipettor with Colored Shaft, Yellow; 20 to 200 µL 2455149 Mfr # 4055
COMPARE
Yellow 200 1
Call For Price
HTL 4056 Discovery Comfort D1000 Single Channel Pipettor with Colored Shaft, Blue; 100 to 1000 µL 2455151 Mfr # 4056
COMPARE
Blue 1000 1
Call For Price
HTL 4057 Discovery Comfort D50 Single Channel Pipettor with Colored Shaft, Yellow; 5 to 50 µL 2455152 Mfr # 4057
COMPARE
Yellow 50 1
Call For Price
HTL 4058 Discovery Comfort D5000 Single Channel Pipettor with Colored Shaft, White; 1 to 5 mL 2455153 Mfr # 4058
COMPARE
White 5 1
Call For Price
HTL 4059 Discovery Comfort D10000 Single Channel Pipettor with Colored Shaft, White; 1 to 10 mL 2455154 Mfr # 4059
COMPARE
White 10 1
Call For Price
HTL 5121 Discovery Comfort DV8-10 Multichannel Pipettor with Gray Shaft, Red, 8-Channel; 1 to 10 µL 2455163 Mfr # 5121
COMPARE
Red 10 8
Call For Price
Showing 1 - 20 of 27