HTL 4042 Discovery Comfort DV10 Single Channel Pipettor with Gray Shaft, Red; 0.5 to 10 µL
Representative image only.
HTL 4042 Discovery Comfort DV10 Single Channel Pipettor with Gray Shaft, Red; 0.5 to 10 µL
| Mfr: 4042| Item: 2455137
Comfort, safety, precision and value make this variable-volume pipette series quite a discovery
Call For Price
Call For Price
Product Description
Key Features
  • Volume locking prevents accidental counter shift
  • Ergonomic design, low pipetting forces help curb repetitive stress injury
  • Fully autoclavable
  • Single-, 8- and 12-channel versions available
  • 3 year warranty
More About this Item
Meticulously designed to ensure your comfort, this UV-resistant pipettor is easy-to-use and calibrate. Discover a 360° rotating manifold, tip ejection with minimal effort, perfect tip sealing and excellent precision. Multichannel models feature a revolutionary suspension system which allows the shafts to retract slightly when they are pressed against a row of pipette tips.
3 Year Warranty
Specifications
  • SterileNo
  • Max Volume (µL)10
  • AutoclavableYes
  • Min Volume (µL)0.5
  • Accuracy (%)0.5
  • Precision (%)0.4
  • Number Of Channels1
  • Tip EjectorYes
  • Color CodeRed
  • DescriptionDiscovery Comfort DV10 Single Channel Pipettor with Gray Shaft, Red; 0.5 to 10 µL