H-B Instrument Enviro-Safe FRIO-Temp Nonhazardous PFA-Coated Verification Bottle Thermometers, Celsius-scale
Enclosed bottle ensures accuracy
  • Enviro-Safe is certified as 100% non-toxic, non-hazardous, biodegradable, and “green”
  • Replace mercury thermometers with the same accuracy without any hazards
  • Statement of accuracy included
Accurately monitor temperatures in freezers, refrigerators, incubators, ovens, and other environments where stability is required. All Thermometers are filled with Enviro-Safe liquid and supplied with a unbreakable plastic bottle filled with Enviro-Safe liquid or glass beads. The thermometer can be removed from the bottle for recalibration if needed. Use the magnetic or hook-and-loop attachment strip to securely attach the bottle to object such as a metal wall or door. All Enviro-Safe™ glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. Each thermometer features a green liquid and white back glass. These thermometers also feature recyclable packaging, RoHS compliant, individual serial numbered and have a statement of accuracy.
Call For Price
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Overall Length (mm)
Availability Pricing
H-B Instrument B60200-0200 FRIO-Temp Incubator Verification Thermometer with Individual Calibration Report; 15 to 30C WW-90300-53 Mfr # 602000200
COMPARE
30 15 180
Call For Price
H-B Instrument B60200-0400 FRIO-Temp Oven Verification Thermometer with Individual Calibration Report; 95 to 115C WW-90300-54 Mfr # 602000400
COMPARE
115 95 200
Call For Price
H-B Instrument B60200-0500 FRIO-Temp Ultra Low Freezer Verification Thermometer with Individual Calibration Report; -90 to 25C WW-90300-55 Mfr # 602000500
COMPARE
25 -90 145
Call For Price
H-B Instrument B60200-0600 FRIO-Temp Refrigerator Verification Thermometer with Individual Calibration Report; -5 to 15C WW-90300-56 Mfr # 602000600
COMPARE
15 -5 125
Call For Price
H-B Instrument B60200-0700 FRIO-Temp Freezer Verification Thermometer with Individual Calibration Report; -30 to 1C WW-90300-57 Mfr # 602000700
COMPARE
1 -30 135
Call For Price
H-B Instrument B60200-0800 FRIO-Temp Blood Bank Verification Thermometer with Individual Calibration Report; -5 to 20C WW-90300-58 Mfr # 602000800
COMPARE
20 -5 135
Call For Price
H-B Instrument B60200-0900 FRIO-Temp Incubator Verification Thermometer with Individual Calibration Report; 15 to 50C WW-90300-59 Mfr # 602000900
COMPARE
50 15 135
Call For Price
H-B Instrument B60200-0000 FRIO-Temp Freezer Verification Thermometer with Individual Calibration Report; -25 to -5C WW-90310-00 Mfr # 602000000
COMPARE
-5 -25 210
Call For Price
H-B Instrument B60200-0100 FRIO-Temp Refrigerator Verification Thermometer with Individual Calibration Report; -2 to 10C WW-90310-01 Mfr # 602000100
COMPARE
10 -2 180
Call For Price
Showing 1- 9 of 9