H-B Instrument B60200-0400 FRIO-Temp Oven Verification Thermometer with Individual Calibration Report; 95 to 115C

Representative image only.

H-B Instrument B60200-0400 FRIO-Temp Oven Verification Thermometer with Individual Calibration Report; 95 to 115C

H-B Instrument | Mfr: B602000400| Item: 9030054

Enclosed bottle ensures accuracy

Call For Price

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

Call For Price

Buy It Together

Current Item

H-B Instrument B60200-0400 FRIO-Temp Oven Verification Thermometer with Individual Calibration Report; 95 to 115C
H-B Instrument B60200-0400 FRIO-Temp Oven Verification Thermometer with Individual Calibration Report; 95 to 115C
9030054

Call For Price
+ =
Total Price 33100.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Enviro-Safe is certified as 100% non-toxic, non-hazardous, biodegradable, and “green”
  • Replace mercury thermometers with the same accuracy without any hazards
  • Statement of accuracy included
  • 5 year warranty

More About this Item

Accurately monitor temperatures in freezers, refrigerators, incubators, ovens, and other environments where stability is required. All Thermometers are filled with Enviro-Safe liquid and supplied with a unbreakable plastic bottle filled with Enviro-Safe liquid or glass beads. The thermometer can be removed from the bottle for recalibration if needed. Use the magnetic or hook-and-loop attachment strip to securely attach the bottle to object such as a metal wall or door. All Enviro-Safe™ glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. Each thermometer features a green liquid and white back glass. These thermometers also feature recyclable packaging, RoHS compliant, individual serial numbered and have a statement of accuracy.

5 Year Warranty
Specifications
  • Length (")7 29/32
  • Min Temperature (° C)95
  • Max Temperature (° C)115
  • Accuracy±0.4°C
  • Division (° C)0.1
  • Thermometer FillBlack Spirit
  • Bottle FillGlass Beads
  • Overall Length (InchShort)7.9
  • Overall Length (mm)200
  • Stem MaterialPFA-Coated White-Back Glass
  • Calibration Points100, 110°C
  • DescriptionFRIO-Temp Oven Verification Thermometer with Individual Calibration Report; 95 to 115C
  • Warranty5 years
Reviews
Q&A