H-B Instrument B60200-0100 FRIO-Temp Refrigerator Verification Thermometer with Individual Calibration Report; -2 to 10C

Representative image only.

H-B Instrument B60200-0100 FRIO-Temp Refrigerator Verification Thermometer with Individual Calibration Report; -2 to 10C

H-B Instrument | Mfr: B602000100| Item: 9031001

Enclosed bottle ensures accuracy

Call For Price

Call For Price
Product Description

Key Features

  • Enviro-Safe is certified as 100% non-toxic, non-hazardous, biodegradable, and “green”
  • Replace mercury thermometers with the same accuracy without any hazards
  • Statement of accuracy included
  • 1 year warranty

More About this Item

Accurately monitor temperatures in freezers, refrigerators, incubators, ovens, and other environments where stability is required. All Thermometers are filled with Enviro-Safe liquid and supplied with a unbreakable plastic bottle filled with Enviro-Safe liquid or glass beads. The thermometer can be removed from the bottle for recalibration if needed. Use the magnetic or hook-and-loop attachment strip to securely attach the bottle to object such as a metal wall or door. All Enviro-Safe™ glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. Each thermometer features a green liquid and white back glass. These thermometers also feature recyclable packaging, RoHS compliant, individual serial numbered and have a statement of accuracy.

1 Year Warranty
Specifications
  • Length (")7 1/10
  • Min Temperature (° C)-2
  • Max Temperature (° C)10
  • Accuracy±0.2°C
  • Division (° C)0.1
  • Thermometer FillBlack Spirit
  • Bottle FillEnviro-Safe® Clear Liquid
  • Overall Length (InchShort)7.1
  • Overall Length (mm)180
  • Stem MaterialPFA-Coated White-Back Glass
  • Calibration Points0, 5°C
  • DescriptionFRIO-Temp Refrigerator Verification Thermometer with Individual Calibration Report; -2 to 10C
  • Warranty1 year
Reviews
Q&A