Fowler Xtra-Value Cal Electronic Calipers

No more converting readings
  • Accuracy: 0.001"/0.02" mm
  • Stainless steel frame
  • Large easy-to-read display
  • Resolution: 0.0005"/0.01 mm
  • Absolute/incremental measurement
  • 4-way measuring
Call For Price

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Max range (in) Resolution
Availability Pricing
Fowler 54-101-150-C Xtra-Value CAL Electronic Caliper, 6"/150 mm; with NIST-Traceable Certificate WW-09925-16 Mfr # 54101150C BOM
COMPARE
±0.001 in 6 0.0005 in
Call For Price
Fowler 54-101-600-1 Xtra-Value CAL Electronic Caliper, Large Display, 6"/150 mm WW-10508-62 Mfr # 54-101-600-1
COMPARE
±0.001 in 6 0.0005 in
Call For Price
Fowler 54-101-150-2 Xtra-Value CAL Electronic Caliper, 6"/150 mm WW-97231-34 Mfr # 54-101-150-2
COMPARE
±0.001 in 6 0.0005 in
Call For Price
Fowler 54-101-300-1 Xtra-Value CAL Electronic Caliper, 12"/300 mm WW-97231-35 Mfr # 54-101-300-1
COMPARE
±0.001 in 12 0.0005 in
Call For Price
Fowler 54-101-900-1 Xtra-Value CAL Electronic Caliper, Large Display, 12"/300 mm WW-97231-36 Mfr # 54-101-900-1
COMPARE
±0.001 in 12 0.0005 in
Call For Price
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat