Fluorinated Jerricans
Constructed of fluorinated HDPE; 53-mm (53B) closures are made of FLPP. Fluorocarbon surface provides an improved barrier and reduces solvent absorption and penetration. Long integral spout allows easy pouring. Includes molded-in liter and gallon graduations.
30,800.00 - ₹43,900.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Cap / Stopper Size Material
Availability Pricing
Thermo Scientific Nalgene 2242-0025 fluorinated jerrican with handle, 10 L 6250100 Mfr # 2242-0025
COMPARE
10 53 mm Fluorinated High Density Polyethylene (FLPE)
₹30,800.00INR / Each
Thermo Scientific Nalgene 2242-0050 fluorinated jerrican with handle, 20 L 6250110 Mfr # 2242-0050
COMPARE
20 53 mm Fluorinated High Density Polyethylene (FLPE)
₹43,900.00INR / Each