Fluke® Pressure Control Modules for 729 Pro Automatic Pressure Calibrators
Interchangeable pressure control modules offer convenience while saving you money
  • Clearly marked maximum pressure range
Switch the pressure control module to match the pressure output range without having to buy a new calibrator.
1,009,000.00 - ₹1,766,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Max Pressure Pressure Resolution Description
Availability Pricing
Fluke FLK-PMM-200K Pressure Control Module, –100 to 200 kPa (–15 to 30 psi) 1778490 Mfr # FLK-PMM-200K
COMPARE
30 0.0001 Pressure Control Module, –100 to 200 kPa (–15 to 30 psi)
₹1,009,000.00INR / Each
Fluke FLK-PMM-200KA Pressure Control Module, 0 to 200 kPa (0 to 30 psi) Absolute 1778491 Mfr # FLK-PMM-200KA
COMPARE
30 0.0001 Pressure Control Module, 0 to 200 kPa (0 to 30 psi) Absolute
₹1,009,000.00INR / Each
Fluke FLK-PMM-2000K Pressure Control Module, –90 to 2000 kPa (–13 to 300 psi) 1778492 Mfr # FLK-PMM-2000K
COMPARE
300 0.001 Pressure Control Module, –90 to 2000 kPa (–13 to 300 psi)
₹1,009,000.00INR / Each
Fluke FLK-PMM-4000K Pressure Control Module, –90 to 4000 kPa (–13 to 600 psi) 1778493 Mfr # FLK-PMM-4000K
COMPARE
600 0.01 Pressure Control Module, –90 to 4000 kPa (–13 to 600 psi)
₹1,261,000.00INR / Each
Fluke FLK-PMM-7000K Pressure Control Module, –90 to 7000 kPa (–13 to 1000 psi) 1778494 Mfr # FLK-PMM-7000K
COMPARE
1000 0.01 Pressure Control Module, –90 to 7000 kPa (–13 to 1000 psi)
₹1,766,000.00INR / Each
Fluke FLK-PMM-7000KA Pressure Control Module, 0 to 7000 kPa (0 to 1000 psi) Absolute 1778495 Mfr # FLK-PMM-7000KA
COMPARE
1000 0.01 Pressure Control Module, 0 to 7000 kPa (0 to 1000 psi) Absolute
₹1,311,000.00INR / Each