Fluke FLK-PMM-7000KA Pressure Control Module, 0 to 7000 kPa (0 to 1000 psi) Absolute
Representative image only.
Fluke FLK-PMM-7000KA Pressure Control Module, 0 to 7000 kPa (0 to 1000 psi) Absolute
Fluke | Mfr: FLK-PMM-7000KA| Item: 1778495
Interchangeable pressure control modules offer convenience while saving you money
₹1,311,000.00INR / Each
₹1,311,000.00INR / Each
Product Description
Key Features
  • Clearly marked maximum pressure range
  • 3 year warranty
More About this Item
Switch the pressure control module to match the pressure output range without having to buy a new calibrator.
Conformite Europeenne3 Year Warranty
Specifications
  • Max Pressure1000
  • Min Pressure0
  • Pressure Resolution0.01
  • Pressure Accuracy0.02% FS
  • DescriptionPressure Control Module, 0 to 7000 kPa (0 to 1000 psi) Absolute
  • Warranty3 years