Fluke FLK-PMM-200K Pressure Control Module, –100 to 200 kPa (–15 to 30 psi)
Representative image only.
Fluke FLK-PMM-200K Pressure Control Module, –100 to 200 kPa (–15 to 30 psi)
Fluke | Mfr: FLK-PMM-200K| Item: 1778490
Interchangeable pressure control modules offer convenience while saving you money
₹1,009,000.00INR / Each
₹1,009,000.00INR / Each
Product Description
Key Features
  • Clearly marked maximum pressure range
  • 3 year warranty
More About this Item
Switch the pressure control module to match the pressure output range without having to buy a new calibrator.
Conformite Europeenne3 Year Warranty
Specifications
  • Max Pressure30
  • Min Pressure-15
  • Pressure Resolution0.0001
  • Pressure Accuracy0.02% FS
  • DescriptionPressure Control Module, –100 to 200 kPa (–15 to 30 psi)
  • Warranty3 years