Fabric Cleanroom Swabs
Eliminates adhesive contamination
  • Ideal for use on sensitive surfaces
  • Enhanced absorbency
  • Excellent chemical resistant
  • Cleanroom processed
  • Autoclavable in dry heat and steam
Fabric Swabs have excellent chemical compatibility. Sealed swabs contain low chloride and other ionic contaminants. Double layer of nonwoven polyester fabric provides enhanced absorbency. The 100% virgin polypropylene handle ensures no additional contaminants are introduced.
7,100.00 - ₹18,700.00INR / Bag of 100
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compliance Swab Total Length (InchShort) Tip Length (InchShort)
Availability Pricing
Texwipe TX759B Cleanroom Swab, 0.39"L Knit Polyester Tip, PP Handle; 500/Pk 3367601 Mfr # TX759B
COMPARE
None 2 3/4 25/64
₹18,200.00INR / Bag of 500
Texwipe TX762 Cleanroom Swab, 0.66"L Knit Polyester Tip, PP Handle; 100/Pk 3367607 Mfr # TX762
COMPARE
None 6 25/64 21/32
₹7,100.00INR / Bag of 100
Texwipe TX743B Cleanroom Swab, 0.472"L Knit Polyester Tip, PP Handle; 500/Pk 3367758 Mfr # TX743B
COMPARE
Class 3 2 3/4 15/32
₹18,700.00INR / Bag of 500
Texwipe TX761 Cleanroom Swab, 0.27"L Knit Polyester Tip, PP Handle; 100/Pk 3367760 Mfr # TX761
COMPARE
Class 3 6 25/64 21/32
₹8,400.00INR / Bag of 100
Texwipe TX714A Cleanroom Swab, 0.5"L Knit Polyester Tip, PP Handle; 100/Pk 3367762 Mfr # TX714A
COMPARE
Class 3 5 1/50 1 1/75
₹12,600.00INR / Bag of 100
Texwipe TX715 Sampling Swab, 0.5"L Knit Polyester Tip, PP Handle; 100/Pk 3367763 Mfr # TX715
COMPARE
None 5 1/50 1 1/75
₹14,400.00INR / Bag of 100
Texwipe TX716 Sampling Swab, 0.5"L Polyester Tip, PP Handle; 20/Pk 3367764 Mfr # TX716
COMPARE
None 5 1/50 1 1/75
₹7,700.00INR / Bag of 20
Texwipe TX714K Sampling Swab, 0.5"L Polyester Tip, PP Handle; 20/Pk 3367765 Mfr # TX714K
COMPARE
None 5 1/50 1 1/75
₹9,900.00INR / Bag of 20
Showing 1- 8 of 8