Extech Refractometers
Includes calibration solution to ensure accurate results
Use these refractometers to take accurate and repeatable measurements of sugar, salt, lubricants, antifreeze, cutting fluids, and more! Simply place 2 or 3 drops of your sample on the prism; the focus adjustment makes the scale easy to read. Automatic temperature compensation ensures accurate readings from 50 to 86°F (10 to 30°C).
35,700.00 - ₹45,900.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Battery Type Max Temperature Compensation (° F) Resolution
Availability Pricing
Extech RF11 Handheld Brix Refractometer with ATC; 0 to 10% 2604802 Mfr # RF11
COMPARE
86 0.1% (Brix)
₹45,900.00INR / Each
Extech RF12 Handheld Brix Refractometer with ATC; 0 to 18% 2604803 Mfr # RF12
COMPARE
86 0.2% (Brix)
₹45,900.00INR / Each
Extech RF20 Refractometer, 100 ppt salt with ATC, 1.000 to 1.070 Refractive Index 2604805 Mfr # RF20
COMPARE
Salt 86 0.001
₹35,700.00INR / Each
Extech RF40 Coolant/Battery Acid Refractometer, -60 to 32 F, 1.15 to 1.30 SG 2604806 Mfr # RF40
COMPARE
Coolant 86 0.01 s.g.