Extech RF40 Coolant/Battery Acid Refractometer, -60 to 32 F, 1.15 to 1.30 SG
Representative image only.
Extech RF40 Coolant/Battery Acid Refractometer, -60 to 32 F, 1.15 to 1.30 SG
Extech | Mfr: RF40| Item: 2604806
Includes calibration solution to ensure accurate results
Call For Price
Call For Price
Buy It Together
Current Item
Extech RF40 Coolant/Battery Acid Refractometer, -60 to 32 F, 1.15 to 1.30 SG
Extech RF40 Coolant/Battery Acid Refractometer, -60 to 32 F, 1.15 to 1.30 SG
2604806
Call For Price
+ =
Total Price 119000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
calibration solution, calibration screwdriver, and case.
Key Features
  • 1 year warranty
More About this Item
Use these refractometers to take accurate and repeatable measurements of sugar, salt, lubricants, antifreeze, cutting fluids, and more! Simply place 2 or 3 drops of your sample on the prism; the focus adjustment makes the scale easy to read. Automatic temperature compensation ensures accurate readings from 50 to 86°F (10 to 30°C).
1 Year Warranty
Specifications
  • Min Battery Specific Gravity1.15
  • Max Battery Specific Gravity1.3
  • Min Ethylene Glycol (° F)-50
  • Max Ethylene Glycol (° F)32
  • Min Propylene Glycol (° F)-60
  • Max Propylene Glycol (° F)32
  • Min Ethylene Glycol (° C)-51
  • Max Ethylene Glycol (° C)0
  • Min Propylene Glycol (° C)-46
  • Max Propylene Glycol (° C)0
  • Resolution0.01 s.g.
  • Temperature Resolution2°F
  • Temperature Accuracy±2%
  • Max Temperature Compensation (° F)86
  • Min Temperature Compensation (° F)50
  • Max Temperature Compensation (° C)30
  • Min Temperature Compensation (° C)10
  • Battery TypeCoolant
  • DescriptionCoolant/Battery Acid Refractometer, -60 to 32 F, 1.15 to 1.30 SG