Cole-Parmer® Digital Refractometers
Never wait for readings again
  • Rugged design is IP65 rated
  • Easy calibration for precise results at all times
  • No more worrying about temperature fluctuations with built-in ATC
These digital refractometers feature an LCD, eliminating reading errors with common handheld refractometers. All models feature built-in automatic temperature compensation (ATC) which removes temperature fluctuations allowing the user to concentrate on the measurements. The compensation range is 41 to 104°F (5 to 40°C).
Many range options are available for measurements within any application. All models also feature a zero% adjust with distilled water, allowing for immediate and easy calibration for precise measurements at all times. The battery life for these units is ≥5000 readings.
65,200.00 - ₹134,000.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Resolution Description
Availability Pricing
COMPARE
±0.1 (Brix), ±0.0003 (RI) 0.1 (Brix), 0.0001 (RI) Digital Refractometer, 1.3330 to 1.4098 RI, 0 - 45%, Brix
₹87,300.00INR / Each
COMPARE
±0.1 (salinity), ±0.0003 (RI) 0.1 (salinity), 0.0001 (RI) Digital Refractometer, 1.3330 to 1.4100 RI , 0 - 28% NaCl
₹103,000.00INR / Each
COMPARE
±0.2 (serum protein), ±0.0003 (RI) 0.2 (serum protein), 0.0001 (RI) Digital Refractometer, 1.3330 to 1.3900 RI , 0 - 14 g/dl Serum Protein
₹65,200.00INR / Each
COMPARE
±0.3 (ethyl alchohol) 0.1 (ethyl alchohol) Digital Refractometer, 0 - 60% v/v Ethanol, 0 - 60%w/w Ethanol
₹89,100.00INR / Each
COMPARE
±0.5 (isopropyl alcohol) 0.1 (isopropyl alcohol) Digital Refractometer, 0 - 40%v/v IPA, 0 - 40%w/w, IPA
₹95,900.00INR / Each
COMPARE
±0.3 (ethylene glycol), ±1°F, °±0.5°C 0.1 (ethylene glycol) Digital Refractometer, 0 to 75% ethylene glycol, –58 to 32°F, –50 to 0°C
₹69,500.00INR / Each
COMPARE
±0.5 (Brix), ±0.0003 (RI) 0.1 (Brix), 0.0001 (RI) Digital Refractometer, 0 - 85% Brix, 1.3330 - 1.5100 RI
₹119,000.00INR / Each
COMPARE
±0.5 (Brix), ±0.0003 (RI) 0.1 (Brix), 0.0001 (RI) Digital Refractometer, 0 - 95% Brix, 1.3330 - 1.5400 RI
₹134,000.00INR / Each
COMPARE
±0.2 (Brix), ±0.0003 (salinity), ±0.1 (RI) 0.1 (Brix), 0.1 (salinity), 0.0001 (RI) Digital Refractometer, 1.3330 to 1.4100 RI , 0 - 45% Brix , 0 - 28% Salinity
₹102,000.00INR / Each
Showing 1- 9 of 9