Extech RF20 Refractometer, 100 ppt salt with ATC, 1.000 to 1.070 Refractive Index
Representative image only.
Extech RF20 Refractometer, 100 ppt salt with ATC, 1.000 to 1.070 Refractive Index
Extech | Mfr: RF20| Item: 2604805
Includes calibration solution to ensure accurate results
₹35,700.00INR / Each
for an additional
₹28,000.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹35,700.00INR / Each
Buy It Together
Current Item
Extech RF20 Refractometer, 100 ppt salt with ATC, 1.000 to 1.070 Refractive Index
Extech RF20 Refractometer, 100 ppt salt with ATC, 1.000 to 1.070 Refractive Index
2604805
₹35,700.00INR / Each
+
InnoCal NIST-Traceable Calibration; Refractometer
1710500
₹28,000.00INR / Each
+ =
Total Price 197700.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Includes
calibration solution, calibration screwdriver, and case.
Key Features
  • 1 year warranty
More About this Item
Use these refractometers to take accurate and repeatable measurements of sugar, salt, lubricants, antifreeze, cutting fluids, and more! Simply place 2 or 3 drops of your sample on the prism; the focus adjustment makes the scale easy to read. Automatic temperature compensation ensures accurate readings from 50 to 86°F (10 to 30°C).
1 Year Warranty
Specifications
  • Min Refractive Index1
  • Max Refractive Index1.07
  • Resolution0.001
  • Max Temperature Compensation (° F)86
  • Min Temperature Compensation (° F)50
  • Max Temperature Compensation (° C)30
  • Min Temperature Compensation (° C)10
  • Battery TypeSalt
  • DescriptionRefractometer, 100 ppt salt with ATC, 1.000 to 1.070 Refractive Index