Environmental Express® TCLP Filters
Meets EPA method 1311 requirements
  • Binder-free borosilicate glass fiber has porosity limits of 0.7 ìm ± 0.1 ìm
  • Exhibit a 98% particle retention rate
  • Available as acid-washed or standard filters
Standard TCLP Filters are ideal for analyzing semi-volatile and volatile organics, and nonmetal inorganics. These filters are tested for wet burst strength and flow rate.
Acid-Washed TCLP Filters are made specifically for evaluating the mobility of metals. They are prepared in a consistent procedure which reduces blank variability and typically provides lower background metals levels than are achieved with on-site filter preparation. These filters are washed in 1 N HNO3 acid and then rinsed three times with copious amounts of deionized water. Each rinse uses more than one liter of deionized water per filter. The filters are then oven dried at 105°C prior to final conversion.
Each box of acid-washed TCLP filters is supplied with an independent certification of preparation warranting that the filters were prepared by Method 1311 protocols and gives demonstrated background TCLP metals levels.
11,100.00 - ₹59,200.00INR / Pkg of 100
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Description
Availability Pricing
COMPARE
47 TCLP Filters, Acid Wash, 47mm, 0.7µm; 100/Pk
Call For Price
COMPARE
76 TCLP Filters, Acid Wash, 76mm, 0.7µm; 50/Pack
₹15,800.00INR / Pkg of 50
COMPARE
90 TCLP Filters, Acid Wash, 90mm, 0.7µm; 50/Pack
₹16,000.00INR / Pkg of 50
COMPARE
110 TCLP Filters, Acid Wash, 110mm, 0.7µm; 50/Pack
₹29,400.00INR / Pkg of 50
COMPARE
125 TCLP Filters, Acid Wash, 125mm, 0.7µm; 50/Pack
₹39,200.00INR / Pkg of 50
COMPARE
142 TCLP Filters, Acid Wash, 142mm, 0.7µm; 50/Pk
₹39,900.00INR / Pkg of 50
COMPARE
150 TCLP Filters, Acid Wash, 150mm, 0.7µm; 50/Pack
₹53,000.00INR / Pkg of 50
COMPARE
47 TCLP Filters, Standard, 47mm, 0.7µm; 100/Pk
₹11,100.00INR / Pkg of 100
COMPARE
76 TCLP Filters, Standard, 76mm, 0.7µm; 100/Pk
₹15,600.00INR / Pkg of 100
COMPARE
90 TCLP Filters, Standard, 90mm, 0.7µm; 100/Pk
₹18,700.00INR / Pkg of 100
COMPARE
110 TCLP Filters, Standard, 110mm, 0.7µm; 100/Pk
₹35,000.00INR / Pkg of 100
COMPARE
125 TCLP Filters, Standard, 125mm, 0.7µm; 100/Pk
₹42,800.00INR / Pkg of 100
COMPARE
142 TCLP Filters, Standard, 142mm, 0.7µm; 100/Pk
₹46,900.00INR / Pkg of 100
COMPARE
150 TCLP Filters, Standard, 150mm, 0.7µm; 100/Pk
₹59,200.00INR / Pkg of 100
Showing 1- 14 of 14