Environmental Express® StepSaver™ Complete Extraction Systems
Small footprint saves valuable benchtop space
  • Can be daisy-chained together to run multiple samples at one
  • Meets EPA method 1664 requirements for oil and grease extractions
  • Two separate chambers help keep water out of your extract
Constructed out of durable PVC and plastic materials, these compact, single-place extraction stations make oil and grease extractions easy. They are designed to be used for any type of solid-phase extraction (SPE) technique using an extraction disk. Single stations can be daisy-chained together to run multiple samples at once for faster sample throughput.
StepSaver systems allow you to easily and efficiently collect the solvent with eluted oil and grease directly into a collection flask. No need to disassemble or reassemble glassware. As fluid enters the collection flask, it passes through a drying cartridge to remove excess water. At this point it is ready to be evaporated without a separate drying step.
The extraction stations are available in three different configurations: disposable polypropylene (PP) filter funnel with UtraPrep® extraction disk, 47-mm glass filter funnel, and 90-mm glass filter funnel.
391,000.00 - ₹505,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Filter Diameter (mm) Funnel Size (mL) Description
Availability Pricing
COMPARE
47 250 Oil and Grease Extraction Complete System, Disposable Funnel
₹391,000.00INR / Each
COMPARE
47 1000 Oil and Grease Extraction Complete System, Glass Funnel; 47 mm
₹428,000.00INR / Each
COMPARE
90 1000 Oil and Grease Extraction Complete System, Glass Funnel; 90 mm
₹505,000.00INR / Each