Environmental Express® Soil-Cell System Accessories
Save time and money with the Soil-Cell extraction system
  • Reduce amount of solvent that is used compared to traditional methods
  • Quick 30 minute extraction saves time
  • Closed-cell technology prevents contamination
The Soil-Cell system is developed for the extraction of semi-volatile organic compounds in soil samples. The Soil-Cell system meets the requirements of the Performance Based Measurement System established by the EPA and replaces microwave technology with a HotBlock and stainless steel Soil Extraction Cells. Analytical results of a CRM (Certified Reference Material) for PAH/BNA and TPH fell within the required acceptance limits and were very reproducible. Real world soil samples were also extracted using this procedure for PAH/BNA, TPH, and PCB compounds and compared to results achieved using SW846 Method 3545A. Results were comparable for all three groups of compounds.
8,100.00 - ₹139,000.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Environmental Express HotBlock Soil-Cell Complete Extraction Cell, Stainless Steel; Each
₹95,700.00INR / Each
COMPARE
Environmental Express HotBlock Soil-Cell Complete Reference Cell, Stainless Steel; Each
₹107,000.00INR / Each
COMPARE
Environmental Express HotBlock Soil-Cell Extraction Cell Threaded Cap; Each
₹8,100.00INR / Each
COMPARE
Environmental Express HotBlock Soil-Cell PTFE-Encapsulated O-Rings; 5/Pk
₹13,100.00INR / Each
COMPARE
Environmental Express HotBlock Soil-Cell Viton O-Rings; 25/Pk
₹9,000.00INR / Each
COMPARE
Environmental Express HotBlock Soil-Cell Extraction Thermometer; Each
₹14,400.00INR / Each
COMPARE
Environmental Express HotBlock Soil-Cell Extraction Foil Ruture Seal; 100/Pk
₹11,400.00INR / Each
COMPARE
Environmental Express HotBlock Soil-Cell Aluminum Rack Set, 25-Well; 2/Set
₹91,200.00INR / Each
COMPARE
Environmental Express HotBlock Soil-Cell Aluminum Rack Set, 35-Well; 3/Set
₹139,000.00INR / Each
Showing 1- 9 of 9