Environmental Express HotBlock Soil-Cell Aluminum Rack Set, 35-Well; 3/Set
Representative image only.
Environmental Express HotBlock Soil-Cell Aluminum Rack Set, 35-Well; 3/Set
Environmental Express | Item: 3520893
Save time and money with the Soil-Cell extraction system
₹139,000.00INR / Each
₹139,000.00INR / Each
Buy It Together
Current Item
Environmental Express HotBlock Soil-Cell Aluminum Rack Set, 35-Well; 3/Set
Environmental Express HotBlock Soil-Cell Aluminum Rack Set, 35-Well; 3/Set
3520893
₹139,000.00INR / Each
+ + =
Total Price 1260000.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Reduce amount of solvent that is used compared to traditional methods
  • Quick 30 minute extraction saves time
  • Closed-cell technology prevents contamination
  • 3 month warranty
More About this Item
The Soil-Cell system is developed for the extraction of semi-volatile organic compounds in soil samples. The Soil-Cell system meets the requirements of the Performance Based Measurement System established by the EPA and replaces microwave technology with a HotBlock and stainless steel Soil Extraction Cells. Analytical results of a CRM (Certified Reference Material) for PAH/BNA and TPH fell within the required acceptance limits and were very reproducible. Real world soil samples were also extracted using this procedure for PAH/BNA, TPH, and PCB compounds and compared to results achieved using SW846 Method 3545A. Results were comparable for all three groups of compounds.
3 Month Warranty
Specifications
  • MaterialAluminum
  • Compatible WithSoil-Cell System
  • DescriptionEnvironmental Express HotBlock Soil-Cell Aluminum Rack Set, 35-Well; 3/Set