Environmental Express HotBlock® Soil-Cell Complete Extraction Cell, Stainless Steel; Each
Representative image only.
Environmental Express HotBlock® Soil-Cell Complete Extraction Cell, Stainless Steel; Each
Environmental Express | Item: 3520881
Save time and money with the Soil-Cell extraction system
₹95,700.00INR / Each
₹95,700.00INR / Each
Buy It Together
Current Item
Environmental Express HotBlock® Soil-Cell Complete Extraction Cell, Stainless Steel; Each
Environmental Express HotBlock® Soil-Cell Complete Extraction Cell, Stainless Steel; Each
3520881
₹95,700.00INR / Each
+ + + =
Total Price 1224800.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Reduce amount of solvent that is used compared to traditional methods
  • Quick 30 minute extraction saves time
  • Closed-cell technology prevents contamination
  • Uses 30 to 40 mL of solvent — a 10X reduction from more traditional techniques such as sonication or Soxhlet!
  • 3 month warranty
More About this Item
The Soil-Cell system is developed for the extraction of semi-volatile organic compounds in soil samples. The Soil-Cell system meets the requirements of the Performance Based Measurement System established by the EPA and replaces microwave technology with a HotBlock and stainless steel Soil Extraction Cells. Analytical results of a CRM (Certified Reference Material) for PAH/BNA and TPH fell within the required acceptance limits and were very reproducible. Real world soil samples were also extracted using this procedure for PAH/BNA, TPH, and PCB compounds and compared to results achieved using SW846 Method 3545A. Results were comparable for all three groups of compounds.
This is one complete extration cell, and includes one stainless steel body, inner lid, O-ring, and cap.
3 Month Warranty
Specifications
  • MaterialStainless steel
  • Compatible WithSoil-Cell System
  • DescriptionEnvironmental Express HotBlock Soil-Cell Complete Extraction Cell, Stainless Steel; Each