Environmental Express® Round Narrow-Mouth Bottles, HDPE
Stand up to your demands
  • Easy-pour design allows for simple handling
These pre-cleaned round narrow-mouth bottles come in two levels. Level 1 bottles are lot controlled, sealed with custody tape, and include a Certificate of Analysis from an independent laboratory.
Level 3 bottles meet EPA guidelines, but do not ship with a certificate of analysis—ideal for general-use.
2,700.00 - ₹43,500.00INR / Case of 48
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap Material Level Number
Availability Pricing
COMPARE
125 LDPE-lined Polypropylene 1
₹4,300.00INR / Case of 48
COMPARE
125 LDPE-lined Polypropylene 1
₹39,900.00INR / Case of 480
COMPARE
250 LDPE-lined Polypropylene 1
₹3,400.00INR / Case of 24
COMPARE
250 LDPE-lined Polypropylene 1
₹33,900.00INR / Case of 336
COMPARE
250 LDPE-lined Polypropylene 3
₹5,500.00INR / Case of 24
COMPARE
250 LDPE-lined Polypropylene 3
₹32,000.00INR / Case of 336
COMPARE
500 LDPE-lined Polypropylene 1
₹3,100.00INR / Case of 24
COMPARE
500 LDPE-lined Polypropylene 1
₹18,200.00INR / Case of 153
COMPARE
500 LDPE-lined Polypropylene 3
₹25,900.00INR / Case of 153
COMPARE
1000 LDPE-lined Polypropylene 1
₹3,200.00INR / Case of 12
COMPARE
1000 LDPE-lined Polypropylene 1
₹13,600.00INR / Case of 66
COMPARE
1000 LDPE-lined Polypropylene 3
₹2,900.00INR / Case of 12
COMPARE
1000 LDPE-lined Polypropylene 3
₹13,000.00INR / Case of 66
COMPARE
₹23,200.00INR / Case of 6
COMPARE
125 LDPE-lined Polypropylene 1
₹2,700.00INR / Case of 24
COMPARE
250 LDPE-lined Polypropylene 1
₹43,500.00INR / Case of 336
COMPARE
500 LDPE-lined Polypropylene 3
₹3,700.00INR / Case of 108
Showing 1- 17 of 17