Environmental Express BPC1051 Pre-Cleaned Round Narrow-Mouth Jug Bottle, HDPE, Level 1, 2 L; 84/CS
Representative image only.
Environmental Express BPC1051 Pre-Cleaned Round Narrow-Mouth Jug Bottle, HDPE, Level 1, 2 L; 84/CS
Environmental Express | Item: 3520734
Stand up to your demands
₹28,200.00INR / Case of 84
₹28,200.00INR / Case of 84
Product Description
Key Features
  • Easy-pour design allows for simple handling
  • Model number BPC1051
More About this Item
These pre-cleaned round narrow-mouth bottles come in two levels. Level 1 bottles are lot controlled, sealed with custody tape, and include a Certificate of Analysis from an independent laboratory.
Level 3 bottles meet EPA guidelines, but do not ship with a certificate of analysis—ideal for general-use.
Specifications
  • MaterialHigh Density Polyethyene (HDPE)
  • DescriptionPre-Cleaned Round Narrow-Mouth Jug Bottle, HDPE, Level 1, 2 L; 84/CS