Environmental Express® MicroBlock™ Distillation System Components
Replacement components for the MicroBlock systems
  • Reduce chemical waste by 90%
  • Get more accurate distillations
  • Comply with all EPA methodologies for the referenced methods; suitable for cyanide (EPA 335.4), phenol (EPA 420.4) and ammonia (EPA 350.1)
Minimize heat loss and maintain uniform distillations by using specially designed MicroBlock glassware. Cap distillation tubes after distillation to prevent evaporation or contamination.
4,300.00 - ₹267,000.00INR / Each
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
MicroBlock™-V, Block Only, 120 VAC
Call For Price
COMPARE
MicroBlock™-V, Block Only, 240 VAC
Call For Price
COMPARE
Cold Finger, for Cyanide Glassware Set
₹41,400.00INR / Each
COMPARE
Cyanide Glassware, 1 Set
₹187,000.00INR / Each
COMPARE
Cap, for Ammonia/Phenol Glassware Set, 10/PK
₹21,700.00INR / Each
COMPARE
Ammonia/Phenol Glassware, 1 Set
₹214,000.00INR / Each
COMPARE
Reaction Tube, for Ammonia/Phenol Glassware Set
₹32,600.00INR / Each
COMPARE
Distillation Head, for Cyanide Glassware Set
Call For Price
COMPARE
Absorption Impinger For Cyanide
₹62,200.00INR / Each
COMPARE
PTFE-Faced Septa, for Ammonia/Phenol Glassware Set, 20/PK
₹35,800.00INR / Pkg of 20
COMPARE
Distillation Head, for Ammonia/Phenol Glassware Set
Call For Price
COMPARE
Tubing and Fitting Sets, for Ammonia/Phenol Glassware Set, 10/ST
₹267,000.00INR / Each
COMPARE
Silicone Sealing Ring, for Ammonia/Phenol Glassware Set, 10/PK
₹40,700.00INR / Pkg of 10
COMPARE
Long Stem Ammonia Outlet Tube, for Ammonia/Phenol Glassware Set
₹4,300.00INR / Each
COMPARE
Phenol Outlet Tube, for Ammonia/Phenol Glassware Set
₹4,300.00INR / Each
COMPARE
Receiver Tube, for Ammonia/Phenol Glassware Set
₹24,300.00INR / Each
COMPARE
MicroBlock™-R, Block Only, 120 VAC
Call For Price
COMPARE
MicroBlock™-R, Block Only, 240 VAC
Call For Price
Showing 1- 18 of 18