Environmental Express Ammonia/Phenol Glassware, 1 Set
Representative image only.
Environmental Express Ammonia/Phenol Glassware, 1 Set
Environmental Express | Item: 3520962
Replacement components for the MicroBlock systems
₹214,000.00INR / Each
₹214,000.00INR / Each
Product Description
Key Features
  • Reduce chemical waste by 90%
  • Get more accurate distillations
  • Comply with all EPA methodologies for the referenced methods; suitable for cyanide (EPA 335.4), phenol (EPA 420.4) and ammonia (EPA 350.1)
  • 3 month warranty
More About this Item
Minimize heat loss and maintain uniform distillations by using specially designed MicroBlock glassware. Cap distillation tubes after distillation to prevent evaporation or contamination.
Includes cold finger, distillation head, reaction tube, and receiver tube. Tubing and fittings are not included.
3 Month Warranty
Specifications
  • MaterialBorosilicate Glass
  • Compatible WithMicroBlock V and R Systems (35209-54, 35209-74, or 35209-75)
  • DescriptionAmmonia/Phenol Glassware, 1 Set