Environmental Express Cap, for Ammonia/Phenol Glassware Set, 10/PK
Representative image only.
Environmental Express Cap, for Ammonia/Phenol Glassware Set, 10/PK
Environmental Express | Item: 3520961
Replacement components for the MicroBlock systems
₹21,700.00INR / Each
₹21,700.00INR / Each
Product Description
Key Features
  • Reduce chemical waste by 90%
  • Get more accurate distillations
  • Comply with all EPA methodologies for the referenced methods; suitable for cyanide (EPA 335.4), phenol (EPA 420.4) and ammonia (EPA 350.1)
  • 3 month warranty
More About this Item
Minimize heat loss and maintain uniform distillations by using specially designed MicroBlock glassware. Cap distillation tubes after distillation to prevent evaporation or contamination.
3 Month Warranty
Specifications
  • DescriptionCap, for Ammonia/Phenol Glassware Set, 10/PK