Environmental Express® Juice-Style, Square Bottles; HDPE
Store more samples side-by-side
  • Provides a better grip and minimizes dropping
  • Leakproof lid keeps samples inside
These pre-cleaned square shaped bottles let you take advantage of precious shelf space. Two levels are available. Level 1 bottles are lot controlled, sealed with custody tape, and include a Certificate of Analysis from an independent laboratory.
Level 3 bottles meet EPA guidelines, but do not ship with a Certificate — ideal for general-use.
28,600.00 - ₹52,900.00INR / Case of 320
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Level Number Special Features
Availability Pricing
COMPARE
250 1 LDPE-lined Polypropylene Cap
₹46,800.00INR / Case of 320
COMPARE
250 1 Rinsed with Hydrochloric Acid; LDPE-lined Polypropylene Cap
Call For Price
COMPARE
250 1 Rinsed with Nitric Acid; LDPE-lined Polypropylene Cap
₹52,900.00INR / Case of 320
COMPARE
250 3 LDPE-lined Polypropylene Cap
₹42,900.00INR / Case of 320
COMPARE
500 1 LDPE-lined Polypropylene Cap
₹43,600.00INR / Case of 240
COMPARE
500 3 LDPE-lined Polypropylene Cap
₹42,700.00INR / Case of 240
COMPARE
1000 1 LDPE-lined Polypropylene Cap
₹29,800.00INR / Case of 108
COMPARE
1000 3 Open-Top Septa Cap
₹28,600.00INR / Case of 108
Showing 1- 8 of 8