Environmental Express® GhostWipe® Dust Sampling Wipes, Standards, and Templates
Simplify your surface sample collection and preparation for lead and beryllium content
  • Just wipe surface you want to test, digest, and analyze results
  • Wipes breakdown completely and wont clog equipment
  • Complies with ASTM method E 1792 for lead or beryllium in dust
The Ghostwipe completely dissolves during the digestion procedure thus allowing everything into solution. Dependent upon the material wiped, filtration may not be necessary. In addition, the Ghostwipe is more rugged than other traditional paper wipes and will not fall apart during the wiping of rough surfaces.
13,900.00 - ₹88,500.00INR / Pkg of 200
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Test Tests Per Kit Description
Availability Pricing
COMPARE
Surface sampling 250 Dust Sampling Template, 10 x 10 cm; 250/Pk
₹13,900.00INR / Pkg of 250
COMPARE
Surface sampling 100 Dust Sampling Template, 30 x 30 cm; 100/Pk
₹27,500.00INR / Pkg of 100
COMPARE
Surface sampling 200 Moist Wipe, 15 x 15 cm, Individually Sealed; 200/Pk
₹19,100.00INR / Pkg of 200
COMPARE
Surface sampling 500 Moist Wipe, 15 x 15 cm, Individually Sealed; 500/Pk
₹28,900.00INR / Pkg of 500
COMPARE
Surface sampling 1000 Moist Wipe, 15 x 15 cm, Individually Sealed; 1000/Pk
₹35,900.00INR / Pkg of 1000
COMPARE
Surface sampling 100 Dry Wipe, 20 x 25 cm; 100/Pk
₹88,500.00INR / Pkg of 100