Environmental Express GhostWipe® Dust Sampling Template, 30 x 30 cm; 100/Pk
Representative image only.
Environmental Express GhostWipe® Dust Sampling Template, 30 x 30 cm; 100/Pk
Environmental Express | Item: 3520871
Simplify your surface sample collection and preparation for lead and beryllium content
₹27,500.00INR / Pkg of 100
₹27,500.00INR / Pkg of 100
Product Description
Key Features
  • Just wipe surface you want to test, digest, and analyze results
  • Wipes breakdown completely and wont clog equipment
  • Complies with ASTM method E 1792 for lead or beryllium in dust
  • Template provides set surface area to test—remain consistent!
  • 3 month warranty
More About this Item
The Ghostwipe completely dissolves during the digestion procedure thus allowing everything into solution. Dependent upon the material wiped, filtration may not be necessary. In addition, the Ghostwipe is more rugged than other traditional paper wipes and will not fall apart during the wiping of rough surfaces.
3 Month Warranty
Specifications
  • TestSurface sampling
  • Tests Per Kit100
  • DescriptionDust Sampling Template, 30 x 30 cm; 100/Pk