Environmental Express® French Square Glass Bottles
Maximize storage space
  • Include a Certificate of Analysis
  • Square design provides optimal use of space—bottles sit side by side
Bottles are available with PTFE-lined caps or open-top septa closures. Polycone PTFE lined caps provide a tighter seal for use with oil samples.
5,900.00 - ₹50,600.00INR / Case of 12
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap Material Description
Availability Pricing
COMPARE
125 PTFE-lined Polypropylene French Square Glass Bottle, Level 1, Clear, 125 mL, PP Cap; 120/CS
₹39,800.00INR / Case of 12
COMPARE
125 PTFE-lined Polypropylene French Square Glass Bottle, Level 1, Clear, 125 mL, PP Cap; 96/CS
₹47,500.00INR / Case of 96
COMPARE
125 Open-Top/Septa French Square Glass Bottle, Level 1, Clear, 125 mL, Open Top; 120/CS
₹8,100.00INR / Case of 12
COMPARE
500 PTFE-lined Polypropylene French Square Glass Bottle, Level 1, Clear, 500 mL; 40/CS
₹23,600.00INR / Case of 40
COMPARE
125 PTFE-lined polypropylene French Square Glass Bottle, Clear, 125mL; 12/CS
₹5,900.00INR / Case of 12
COMPARE
1000 PTFE-lined Polypropylene French Square Glass Bottle, Level 1, Clear, 1000 mL; 24/CS
₹50,600.00INR / Case of 24
COMPARE
500 PTFE-lined Polypropylene French Square Glass Bottle, Level 3, Clear, 500 mL; 40/CS
₹16,600.00INR / Case of 40
Showing 1- 7 of 7