Environmental Express Disposable Sterile Sampling Vials
Sterile vials are designed for coliform water sampling
  • Comes with all the necessary certificates of sterility
  • Screw-top design provides ease of use
  • Holds a total volume of 120 mL
  • Offers clean view with UV transparency
  • Each vial is shrink-wrapped to ensure sterility
These disposable vials are sterile and ideal for coliform water sampling and sample dichlorination applications. Each vial is marked at the 100 mL fill line. Vials with thiosulfate contain enough sodium thiosulfate to neutralize 15 mg of residual chlorine. When filled, the vials meet the 0.008% sodium thiosulfate concentration requirement for sample collection containers.
18,300.00 - ₹18,500.00INR / Case of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Special Features
Availability Pricing
COMPARE
Vial with Thiosulfate
₹18,500.00INR / Case of 100
COMPARE
Vial without Thiosulfate
₹18,300.00INR / Pkg of 100